Affinity chromatography media, Blue Sepharose™ High Performance
Artikelnr. 90-1001-23
Lieferant: Cytiva
CAS-Nummer:
Spezifikationen
- Particle Size:~34 µm
- Pk:50 ml
- Description:Blue Sepharose™ High Performance
- pH range:4 - 12
- Matrix:Highly cross-linked agarose, 6%
- Ligand:Cibacron Blue F3G-A
- Storage temperature:4 - 30 °C
- Ligand density:4 mg Cibacron Blue F3G-A/ml medium
- Ligand coupling method:Triazine
- Description:Blue Sepharose™ High Performance
- Type:Liquid
Spezifikationen
Über diesen Artikel
This media is suitable for affinity chromatography.
Chemical stability: Commonly used buffers, 70% ethanol, 8 M urea and 6 M guanidine hydrochloride.
Storage solution: 20% ethanol