Orders LinkContactUsLinkComponent
Accessories for circulators
Accessories for circulators
Supplier:   HUBER
Accessories for circulators
Supplier:  HUBER
Certificates
Accessories for circulators
Accessories for circulators
24 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • For
    Availability
    Price
  • HUBE337657
    2-way header, M16×1
    2-way header, M16×1
    The 2-way header and the TCS should be connected with a suitable length of insulated hose. Universal mounting device for fixing in position is included. For direct connection to the unit, an additional elbow connector is required.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 275,00 €
    275,00 €
    1 * 1 ST
       275,00 €
  • HUBE341870
    3-way header, M16×1
    3-way header, M16×1
    The 3-way header and the TCS should be connected with a suitable length of insulated hose. Universal mounting device for fixing in position is included. For direct connection to the unit, an additional elbow connector is required.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 375,00 €
    375,00 €
    1 * 1 ST
       375,00 €
  • HUBE341871
    4-way header, M16×1
    4-way header, M16×1
    The 4-way header with an universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 390,00 €
    390,00 €
    1 * 1 ST
       390,00 €
  • HUBE343295
    3-way header with ball valves, M16×1
    3-way header with ball valves, M16×1
    The 3-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 520,00 €
    520,00 €
    1 * 1 ST
       520,00 €
  • HUBE343304
    4-way header with ball valves, M16×1
    4-way header with ball valves, M16×1
    The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 565,00 €
    565,00 €
    1 * 1 ST
       565,00 €
  • HUBE343230
    2-way header, M30×1,5
    2-way header, M30×1,5
    The 2-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 480,00 €
    480,00 €
    1 * 1 ST
       480,00 €
  • HUBE342639
    3-way header, M30×1,5
    3-way header, M30×1,5
    The 2-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 545,00 €
    545,00 €
    1 * 1 ST
       545,00 €
  • HUBE343314
    2-way header with ball valves, M30×1,5
    2-way header with ball valves, M30×1,5
    The 2-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 760,00 €
    760,00 €
    1 * 1 ST
       760,00 €
  • HUBE343317
    3-way header with ball valves, M30×1,5
    3-way header with ball valves, M30×1,5
    The 3-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 970,00 €
    970,00 €
    1 * 1 ST
       970,00 €
  • HUBE342090
    2-way header, M38×1,5
    2-way header, M38×1,5
    The 2-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 530,00 €
    530,00 €
    1 * 1 ST
       530,00 €
  • HUBE343234
    3-way header, M38×1,5
    3-way header, M38×1,5
    The 3-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 600,00 €
    600,00 €
    1 * 1 ST
       600,00 €
  • HUBE343235
    4-way header, M38×1,5
    4-way header, M38×1,5
    The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 670,00 €
    670,00 €
    1 * 1 ST
       670,00 €
  • HUBE343321
    2-way header with ball valves, M38×1,5
    2-way header with ball valves, M38×1,5
    The 2-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 965,00 €
    965,00 €
    1 * 1 ST
       965,00 €
  • HUBE343329
    3-way header with ball valves, M38×1,5
    3-way header with ball valves, M38×1,5
    The 3-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 280,00 €
    1 280,00 €
    1 * 1 ST
       1 280,00 €
  • HUBE343331
    4-way header with ball valves, M38×1,5
    4-way header with ball valves, M38×1,5
    The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 550,00 €
    1 550,00 €
    1 * 1 ST
       1 550,00 €
  • HUBE341892
    5-way header, M16×1
    5-way header, M16×1
    The 5-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 510,00 €
    510,00 €
    1 * 1 ST
       510,00 €
  • HUBE343305
    5-way header with ball valves, M16×1
    5-way header with ball valves, M16×1
    The 5-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 810,00 €
    810,00 €
    1 * 1 ST
       810,00 €
  • HUBE350025
    2-way valve system, 19 mm
    2-way valve system, 19 mm
    2-way valve system with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 595,00 €
    595,00 €
    1 * 1 ST
       595,00 €
  • HUBE343221
    2-way header, M24×1,5
    2-way header, M24×1,5
    The 2-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 335,00 €
    335,00 €
    1 * 1 ST
       335,00 €
  • HUBE343226
    3-way header, M24×1,5
    3-way header, M24×1,5
    The 3-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HUBE343228
    4-way header, M24×1,5
    4-way header, M24×1,5
    The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 490,00 €
    490,00 €
    1 * 1 ST
       490,00 €
  • HUBE343306
    2-way valve system, M24×1,5
    2-way valve system, M24×1,5
    2-way valve system with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 560,00 €
    560,00 €
    1 * 1 ST
       560,00 €
  • HUBE343308
    3-way header with ball valves, M24×1,5
    3-way header with ball valves, M24×1,5
    3-way valve system with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 845,00 €
    845,00 €
    1 * 1 ST
       845,00 €
  • HUBE343310
    4-way header with ball valves, M24×1,5
    4-way header with ball valves, M24×1,5
    The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. Note: The manifold must not be mounted on the unit.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 930,00 €
    930,00 €
    1 * 1 ST
       930,00 €