ContactUsLinkComponent
Humidity kit with Bluetooth®
Humidity kit with Bluetooth®
Catalog # TEST0563.4404
Supplier:  Testo
undefined
Humidity kit with Bluetooth®
Catalog # TEST0563.4404
Supplier:  Testo
Specifications
  • Interface:
    Bluetooth
  • Description:
    Air velocity and IAQ measuring instrument
  • Seq:
    74
  • Pk:
    1
Specifications
About this item
Humidity kit with Bluetooth is equipped with clearly structured measurement menus for long-term measurements and parallel determination of humidity and air temperature in storage, refrigerated, and work rooms, and in ventilation ducts.
  • Convenience and maximum flexibility for duct and outlet measurements
  • Long-term monitoring of indoor air quality
  • Turbulence measurement in accordance with EN ISO 7730 / ASHRAE 55
Convenient for wireless probes, internal memory for data storage, and USB port for data export. Ideal for different range of probes for cleanrooms and laboratories, for carrying out measurements in storage, refrigeration, and work rooms, in ventilation ducts, and also on refrigeration systems and heat pumps.
Delivery: Supplied with testo 440 air velocity and IAQ measuring instrument,three AA batteries, USB cable, and calibration protocol (TEST0560.4401), humidity/temperature probe (Ø 0.47") with Bluetooth* (consisting of humidity/temperature probe head and Bluetooth handle), four AA batteries, and calibration protocol (TEST0636.9731), basic case for testo 440 and one probe.
Caution: Do not use the probe in condensing atmospheres. For continuous application in high-humidity ranges:
> 80% RH at ≤ 30 °C for > 12 h
> 60% RH at > 30 °C for > 12 h