Orders LinkContactUsLinkComponent
Electrolyte solutions, aqueous
Electrolyte solutions, aqueous
Electrolyte solutions, aqueous
Supplier:  SI Analytics
Certificates
Electrolyte solutions, aqueous
Electrolyte solutions, aqueous
6 Options Available Below
Product Details & Documents
Electrolyte aqueous solutions are used for reference electrodes, as electrolyte bridges and for storage.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • SCOI285138151
    Electrolyte solution, L 200 low temperature electrolyte (–30 °C), 1000 ml in DURAN® glass bottle
    Electrolyte solution, L 200 low temperature electrolyte (–30 °C), 1000 ml in DURAN® glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 149,00 €
    149,00 €
    1 * 1 L
       149,00 €
  • SCOI285138590
    Electrolyte solution, L 911 storage electrolyte solution, sterilized, 1000 ml in DURAN® glass bottle
    Electrolyte solution, L 911 storage electrolyte solution, sterilized, 1000 ml in DURAN® glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 73,00 €
    73,00 €
    1 * 1 L
       73,00 €
  • 662-6184
    Electrolyte solution, L 2114 2 mol/l KNO3 + 0.001 mol/l KCl, for Ag combination electrodes, 250 ml in DURAN® glass bottle
    Electrolyte solution, L 2114 2 mol/l KNO3 + 0.001 mol/l KCl, for Ag combination electrodes, 250 ml in DURAN® glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 36,40 €
    36,40 €
    1 * 250 mL
       36,40 €
  • SCOI285136923
    Electrolyte solution, L 2214 2 mol/l KNO3 + 0.001 mol/l KCl, for Ag combination electrodes, thickened, 250 ml in DURAN® glass bottle
    Electrolyte solution, L 2214 2 mol/l KNO3 + 0.001 mol/l KCl, for Ag combination electrodes, thickened, 250 ml in DURAN® glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 48,30 €
    48,30 €
    1 * 250 mL
       48,30 €
  • SCOI285138440
    Electrolyte solution L3004 cert - KCl 3.0 mol/l, sterilized, 250 ml in DURAN® glass bottle
    Electrolyte solution L3004 cert - KCl 3.0 mol/l, sterilized, 250 ml in DURAN® glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 51,00 €
    51,00 €
    1 * 250 mL
       51,00 €
  • SCOI285138560
    Electrolyte solution, L 9114 storage electrolyte solution, sterilized, 250 ml in DURAN® glass bottle
    Electrolyte solution, L 9114 storage electrolyte solution, sterilized, 250 ml in DURAN® glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 49,70 €
    49,70 €
    1 * 250 mL
       49,70 €