Multi-element noble metals standards, VHG
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
Multi element standards for Au, Ir, Os, Pd, Pt, Re, Rh, Ru at 100 µg/ml in 20% HCl.
Frequently Bought Together
Multi element standards for Au, Ir, Os, Pd, Pt, Re, Rh, Ru at 100 µg/ml in 20% HCl.