ContactUsLinkComponent
Blades for safety knives, No. 86, Styropor
Blades for safety knives, No. 86, Styropor
Catalog # MARO86.60
Supplier:  MARTOR ARGENTAX
undefined
Blades for safety knives, No. 86, Styropor
Catalog # MARO86.60
Supplier:  MARTOR ARGENTAX
Specifications
  • Description:
    Replacement blade, No. 86 Styropor, double bevel edge-ground, 2-sided ground-edge
  • L×W:
    95×8,90 mm
  • Thickness:
    0,40 mm
  • Environmentally Preferable:
  • Pk:
    10
Specifications
About this item
Styropor blade for use with safety knives.
  • 2-sided ground-edge
  • Double bevel edge-ground
Mountable in:
ARGENTAX CUTTEX