Lugol's iodine solution stabilised for Gram staining, Sigma-Aldrich®
: Merck
:
:1.02 g/cm3 (20 °C)
:M-Clarity™ quality level = MQ300
:Stabilised with polyvinylpyrrolidon (PVP)
:Ambient
:1
- Environmentally Preferable:
- Suitability for microscopy:passes test
- gram-positve microorganisms:blue-violet
- gram-negative microorganisms:pink to red
Frequently Bought Together