Orders LinkContactUsLinkComponent
Heat resistant gloves, uvex k-basic extra
Heat resistant gloves, uvex k-basic extra
Supplier:   UVEX
Heat resistant gloves, uvex k-basic extra
Supplier:  UVEX
Certificates
Heat resistant gloves, uvex k-basic extra
3 Options Available Below
Product Details & Documents
These coarse-knitted gloves made of 100% Kevlar® are lined with cotton, making them ideal for protecting the wearer against heat and cuts.
  • Temperature resistant up to +250 °C
  • Excellent protection against cuts
  • Breathable
  • Very good thermal insulation in direct contact with warm to hot objects
  • Good wearer comfort with the cotton lining on the inside
Ideal for welding applications, metal processing, automotive industry, glass industry and foundries.
Certifications: EN 388:2016 (2442D), EN 407 (X2XXXX)
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • UVEX6017902
  • UVEX6017904
  • UVEX6017905
  • Model:
  • 6658
  • 6658
  • 6658
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Cotton
  • Cotton
  • Cotton
  • Gauge:
  • 7
  • 7
  • 7
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (2442D), EN 407 (X2XXXX)
  • EN 388:2016 (2442D), EN 407 (X2XXXX)
  • EN 388:2016 (2442D), EN 407 (X2XXXX)
Specifications
Product Family Options
Product Information
  • Glove size
    Colour
    Length
    Availability
    Price
  • 8
    Yellow
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 32,90 €
    32,90 €
    1 * 1 PAAR
       32,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    UVEX6017902
    Model
    6658
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Cotton
    Gauge
    7
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (2442D), EN 407 (X2XXXX)
  • 10
    Yellow
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 32,90 €
    32,90 €
    1 * 1 PAAR
       32,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    UVEX6017904
    Model
    6658
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Cotton
    Gauge
    7
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (2442D), EN 407 (X2XXXX)
  • 12
    Yellow
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 32,90 €
    32,90 €
    1 * 1 PAAR
       32,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    UVEX6017905
    Model
    6658
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Cotton
    Gauge
    7
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (2442D), EN 407 (X2XXXX)