ContactUsLinkComponent
Reusable earplugs, whisper+ detec
Reusable earplugs, whisper+ detec
Catalog # UVEX2111.239
Supplier:  UVEX
undefined
Reusable earplugs, whisper+ detec
Catalog # UVEX2111.239
Supplier:  UVEX
Specifications
  • Colour:
    Blue
  • Description:
    Earplugs, with cord, in box
  • Size:
    M
  • Pk:
    50 PAIR
Specifications
About this item
Earplugs with cord are conveniently at hand, whenever required. Suitable for use in dirty working environments.
  • Can be worn comfortably around the neck
  • Hygienic storage in an individual carrying case
  • Dirt-repellent surface can be cleaned using soap and water
  • Maintain the wearer's comfort and safety
Suitable for low noise areas.
SNR: 27 dB
Certifications: EN 352-2:2002