ContactUsLinkComponent
Furnace tong, Ward pattern
Furnace tong, Ward pattern
Catalog # RLAA5704
Supplier:  R L ENTERPRISES
Furnace tong, Ward pattern
Catalog # RLAA5704
Supplier:  R L ENTERPRISES
Specifications
  • Length:
    590 mm
  • Pk:
    1
  • Type:
    Furnace tong
Specifications
About this item
This tong is used for protection and safety when handling heated objects in the laboratory.
  • Manufactured from stainless steel
  • For handling a wide variety of crucibles, especially in muffle furnaces
  • Operated by thumb action with one hand
Fitted with a circular shield to protect the hand against heat from the furnace.