Doramapimod ≥99% (by HPLC), Calbiochem®
Catalog # 506172-10MG
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Purity:≥99% by HPLC
- Form:White Solid
Specification Test Results
About this item
:
:527.669 g/mol
Storage Temperature:Refrigerator
CAS Number:285983-48-4
Formula:C₃₁H₃₇N₅O₃
MDL Number:MFCD09752957