Order Entry
Austria
ContactUsLinkComponent
 
di-Sodium L(+)-tartrate dihydrate 99.0-101.0% (by perchloric acid titration) Reag. Ph. Eur.
  APPC121719.1211
 :  
undefined
di-Sodium L(+)-tartrate dihydrate 99.0-101.0% (by perchloric acid titration) Reag. Ph. Eur.
  APPC121719.1211
 :  

 

  • Pk:
    1.000 g
  • Seq:
    3745

 

 

  • Insoluble Matter in H₂O:
    ≤0.005%
  • Chloride (Cl):
    ≤0.0005%
  • Ammonium (NH₄):
    ≤0.003%
  • Phosphate (PO₄):
    ≤0.0005%
  • Sulphate (SO₄):
    ≤0.002%
  • Heavy Metals (as Pb):
    ≤0.0005%
  • As:
    ≤0.00005%
  • Ca:
    ≤0.005%
  • Cu:
    ≤0.0005%
  • Fe:
    ≤0.0005%
  • K:
    ≤0.002%
  • Ni:
    ≤0.0005%
  • PB:
    ≤0.0005%
  • Assay (Perchl. Ac.):
    99.0 - 101.0%
  • pH of 5% solution:
    7.0 - 9.0

 

 

 :
 :230.08 g/mol
 :150 °C
 :Ambient
 :212-773-3
 :6106-24-7
 :NaOOC(CHOH)₂COONa·2H₂O
 :MFCD00150035