


N,N-Dimethylformamide ≥99.9%, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Flash Pt:57.5 °C
UN:2265
ADR:3,III
:M-Clarity™ quality level = MQ100
:GHS02
:GHS08
:GHS07
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Purity (GC):≥ 99.9 %
- Water:≤ 0.03 %
- Transmission (at 275 nm):≥ 65 %
- Transmission (at 300 nm):≥ 90 %
- Transmission (at 350 nm):≥ 99.5 %
- Transmission (at 400 nm):≥ 99.5 %
Specification Test Results
Frequently Bought Together