Order Entry
Austria
ContactUsLinkComponent
Transport Boxes, Fiberboard
Transport Boxes, Fiberboard
Catalog # CORT3CAH000981
Supplier:  E3 Cortex
Transport Boxes, Fiberboard
Catalog # CORT3CAH000981
Supplier:  E3 Cortex

Specifications

  • W×D×H ext.:
    425×425×450 mm
  • Description:
    Transport box for hazardous material, load capacity 31 kg
  • Pk:
    1
  • W×D×H int.:
    400×400×400 mm

Specifications

About this item

These cardboards boxes are used for the transfer of dangerous and\or regulated goods by air, road and maritime traffic. They allow the shipping of liquid products or solids of any classes (except 6.2 and 7) according to regulations in force (IATA, ADR, IMDG, RID).

  • Assembly in two minutes
  • Absorbent vermiculite for dangerous products of the groups of Packaging I, II and III
  • All the references are UN certified 4GV, allowing the use of every type of internal bowl
  • Must be used with an absorbent
Certifications: UN certified.
Packaging: Delivered with plastic bag.