Order Entry
Austria
ContactUsLinkComponent
Bouin liquor
Bouin liquor
Catalog # APPC254102.1611
Supplier:  PanReac AppliChem
CAS Number:  
Bouin liquor
Catalog # APPC254102.1611
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1.000 mL
  • Description:
    Bouin liquor

Specifications

About this item

Fixative for preserving soft and delicate structures, used as a mordant in various trichrome procedures.

Composition:
Picric Acid moistened with ~33% H₂O: 1,125 g
Acetic Acid glacial: 5 ml
Formaldehyde 35 to 40%: 25 ml
Water: 77 ml