OpenLab CDS Software
About this item
Agilent OpenLab CDS is the next generation chromatography data system, designed for ease of use and productivity. OpenLab CDS offers the most comprehensive instrument control and data acquisition for Hitachi HPLC and amino acid analysers. In addition, it supports an expanded array of instrumentation from other vendors, for example Agilent LC, GC and LC/MS. Whether you like to control a single instrument or a huge network of different analytical instruments, OpenLab CDS grows with your needs.
- Direct control of Hitachi HPLC and amino acid analyser
- Workstation and Client-Server functionality with a scalable architecture that allows the system grow with your lab
- One software for LC, GC, GC/MS, LC/MS and other vendors instruments provides same user experience and allows viewing data across techniques and instruments with the same user interface
- New user interface displays only the relevant tools and information you need for your current work
- Built-in custom calculations eliminate errors caused by data export
- Advanced reporting capabilities make generating even complex reports faster and easier
- Superior data analysis such as peak explorer, speed data processing and review
- Centralized instrument management and system administration to increase lab efficiency and instrument uptime
- Built-in technical controls ensure the security of your data, control access and facilitates compliance
- Audit trails now include confirmation and documentation of audit trail reviews as part of the electronic record - so you can review the audit trail and records in parallel
- Automated software migration tools preserve result sets, methods, user and instrument information from legacy ChemStation, ChemStation edition, EZChrom edition, and Galaxie CDS (workstation only)
Including a new 'Custom Calculator', OpenLab CDS allow you to produce new values based on mathematical calculations of system-generated results. Calculations can be made peak-by-peak or across an entire injecting-set, and complex, multi-variable calculations are also possible.
In addition to the powerful peak integration and exploring tools, OpenLab CDS contains a versatile reporting engine that generates standard reports using more than 20 out-of-the-box templates. You can easily modify these templates to suit your lab or generate custom reports for export into common word processing formats.
Certifications: With full FDA 21 CFR Part 11 compliance, OpenLab CDS will satisfy all regulatory requirements. You will get all the capabilities you need to stay compliant and mitigate regulatory exposure.
15 Options Available Below
Product Details & Documents
Agilent OpenLab CDS is the next generation chromatography data system, designed for ease of use and productivity. OpenLab CDS offers the most comprehensive instrument control and data acquisition for Hitachi HPLC and amino acid analysers. In addition, it supports an expanded array of instrumentation from other vendors, for example Agilent LC, GC and LC/MS. Whether you like to control a single instrument or a huge network of different analytical instruments, OpenLab CDS grows with your needs.
- Direct control of Hitachi HPLC and amino acid analyser
- Workstation and Client-Server functionality with a scalable architecture that allows the system grow with your lab
- One software for LC, GC, GC/MS, LC/MS and other vendors instruments provides same user experience and allows viewing data across techniques and instruments with the same user interface
- New user interface displays only the relevant tools and information you need for your current work
- Built-in custom calculations eliminate errors caused by data export
- Advanced reporting capabilities make generating even complex reports faster and easier
- Superior data analysis such as peak explorer, speed data processing and review
- Centralized instrument management and system administration to increase lab efficiency and instrument uptime
- Built-in technical controls ensure the security of your data, control access and facilitates compliance
- Audit trails now include confirmation and documentation of audit trail reviews as part of the electronic record - so you can review the audit trail and records in parallel
- Automated software migration tools preserve result sets, methods, user and instrument information from legacy ChemStation, ChemStation edition, EZChrom edition, and Galaxie CDS (workstation only)
Including a new 'Custom Calculator', OpenLab CDS allow you to produce new values based on mathematical calculations of system-generated results. Calculations can be made peak-by-peak or across an entire injecting-set, and complex, multi-variable calculations are also possible.
In addition to the powerful peak integration and exploring tools, OpenLab CDS contains a versatile reporting engine that generates standard reports using more than 20 out-of-the-box templates. You can easily modify these templates to suit your lab or generate custom reports for export into common word processing formats.
Certifications: With full FDA 21 CFR Part 11 compliance, OpenLab CDS will satisfy all regulatory requirements. You will get all the capabilities you need to stay compliant and mitigate regulatory exposure.
Documents
Agilent Technologies Software, OpenLAB 2 CDS
Product Literature
Product Family Options
Workstation software
Product Information
- AvailabilityPrice
- 906-0103OpenLab CDS standard workstation software with core software and 1 year SMA, no system connection includedOpenLab CDS standard workstation software with core software and 1 year SMA, no system connection includedLog in for availability5 420,00 €
- 906-0104OpenLab CDS PLUS workstation software with core software, 1 year SMA and integrated content managment system, no system connection includedOpenLab CDS PLUS workstation software with core software, 1 year SMA and integrated content managment system, no system connection includedLog in for availability8 530,00 €
- SCSOK4626BAOpenLab CDS WS Plus workstation software with core softwareOpenLab CDS WS Plus workstation software with core softwareLog in for availability1 * 1 ST9 890,00 €
- 906-0109OpenLab CDS Hitachi LC control license only, instrument control driverOpenLab CDS Hitachi LC control license only, instrument control driverLog in for availability4 700,00 €
Additional instrument controls
Product Information
- AvailabilityPrice
- 906-0106OpenLab CDS instrument control driver for connection of a Agilent quadrupole gas chromatography/mass spectrometry system or a liquid chromatography/mass spectrometry systemOpenLab CDS instrument control driver for connection of a Agilent quadrupole gas chromatography/mass spectrometry system or a liquid chromatography/mass spectrometry systemLog in for availability2 640,00 €
- 906-0105OpenLab CDS instrument control driver for connection of Agilent LC, GC, A/D, UV or 3D-DADOpenLab CDS instrument control driver for connection of Agilent LC, GC, A/D, UV or 3D-DADLog in for availability1 330,00 €
- 906-0112OpenLab CDS shimadzu GC control license only, instrument control driverOpenLab CDS shimadzu GC control license only, instrument control driverLog in for availability4 520,00 €
- 906-0113OpenLab CDS shimadzu LC control license only, instrument control driverOpenLab CDS shimadzu LC control license only, instrument control driverLog in for availability4 520,00 €
- 906-0111OpenLab CDS thermo LC control license only, instrument control driverOpenLab CDS thermo LC control license only, instrument control driverLog in for availability4 520,00 €
- 906-0115OpenLab CDS valco valve control license only, instrument control driverOpenLab CDS valco valve control license only, instrument control driverLog in for availability1 260,00 €
- 906-0114OpenLab CDS varian-bruker GC control license only, instrument control driverOpenLab CDS varian-bruker GC control license only, instrument control driverLog in for availability4 520,00 €
- 906-0110OpenLab CDS waters LC control license only, instrument control driverOpenLab CDS waters LC control license only, instrument control driverLog in for availability4 520,00 €
Network software
Product Information
- AvailabilityPrice
- 906-0107OpenLab CDS AIC software (AIC= Agilent Instrument Controller); kit includes software and license for one AICOpenLab CDS AIC software (AIC= Agilent Instrument Controller); kit includes software and license for one AICLog in for availability5 020,00 €
- SCSOK4630BAOpenLab CDS shared services server software and license
Upgrade
Product Information