ContactUsLinkComponent
UV transilluminator bulbs for myECL™ imager
UV transilluminator bulbs for myECL™ imager
Catalog # PIER62255
UV transilluminator bulbs for myECL™ imager
Catalog # PIER62255
Specifications
  • Description:
    UV transilluminator bulb, 365 nm
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
These lamp bulbs with ultraviolet emission are used to replace the one of six pre-installed 306 nm UV bulbs in the myECL™ imager. All six lamps must be the same bulb type (365 or 306 nm).