Rapid resolution high definition (RRHD) columns, ZORBAX® 300SB
Supplier: Agilent
Certificates
About this item
300 stablebond columns are designed for optimal use at low pH.
At pH 6 to 8, highest column stability for all silica-based columns is obtained by operating at temperatures of <40 °C and using low buffer concentrations in the range of 0,01 to 0,02 M.
Specifications
- Catalog No:
- HEWL863750-902
- HEWL858750-944
- HEWL965600-944
- HEWL858750-902
- HEWL857750-906
- HEWL857750-902
- HEWL858750-906
- HEWL857750-944
- HEWL858750-909
- HEWL857750-901
- HEWL858750-901
- HEWL857750-909
- Description:
- 300SB-C18
- 300-Diphenyl
- 300-Diphenyl
- 300SB-C18
- 300SB-C8
- 300SB-C18
- 300SB-C8
- 300-Diphenyl
- 300SB-C3
- 300SB-C3
- Carbon load:
- 2,8%
- 1,9%
- 1,9%
- 2,8%
- 1,5%
- 2,8%
- 1,5%
- 1,9%
- 1,1%
- 1,1%
- Endcapped:
- No
- Yes
- Yes
- No
- No
- No
- No
- Yes
- No
- No
- pH range:
- 1,0 - 8,0
- 1,0 - 8,0
- 1,0 - 8,0
- 1,0 - 8,0
- 1,0 - 8,0
- 1,0 - 8,0
- 1,0 - 8,0
- 1,0 - 8,0
- 1,0 - 8,0
- 1,0 - 8,0
- Surface area:
- 45 m²/g
- 45 m²/g
- 45 m²/g
- 45 m²/g
- 45 m²/g
- 45 m²/g
- 45 m²/g
- 45 m²/g
- 45 m²/g
- 45 m²/g
- Temperature limit:
- 90 °C
- 80 °C
- 80 °C
- 90 °C
- 80 °C
- 90 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP code:
- L1
- L11
- L11
- L1
- L56
- L1
- L56
- L11
- L7
- L7
Specifications
Frequently Bought Together