Order Entry
Austria
ContactUsLinkComponent
2,4,6-Trinitrobenzenesulphonic acid (TNBS) 5% (w/v) in water, BioReagent suitable for determination of primary amines, Sigma-Aldrich®
2,4,6-Trinitrobenzenesulphonic acid (TNBS) 5% (w/v) in water, BioReagent suitable for determination of primary amines, Sigma-Aldrich®
Supplier:  Merck
Certificates
2,4,6-Trinitrobenzenesulphonic acid (TNBS) 5% (w/v) in water, BioReagent suitable for determination of primary amines, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance (Color):
    Yellow
  • Appearance (Turbidity):
    Clear to Slightly Hazy
  • Appearance (Form):
    Liquid
  • Concentration (w/v):
    4.8 - 5.2 %

Specification Test Results

Related Information

Frequently Bought Together