Order Entry
Austria
Orders LinkContactUsLinkComponent
Forceps, PFA

Forceps, PFA

Straight, rounded tip, 1,5 mm tip size, White, 127 mm
Catalog # ENTEC20-0215
Supplier:   ENTEGRIS
Forceps, PFA
Catalog # ENTEC20-0215
Supplier:  ENTEGRIS
Forceps, PFA

Straight, rounded tip, 1,5 mm tip size, White, 127 mm

Price
Inquire For Price
Log in for availability
1 * 4 ST
Product Details & Documents

PFA, white.

  • Straight forceps for substrate handling
  • Extra heavy for much stronger grip at ends

Packaging: Packaged in protective cases.

Specifications
  • Type:
    Straight, rounded tip, 1,5 mm tip size
  • Colour:
    White
  • Length:
    127 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.