Organic reference standard, Methoxychlor, Dr Ehrenstorfer
EHERL15060000IO
:
- Concentration:10 µg/ml in isooctane
- Pk:10 mL
- Standards:Reference standard
- CAS No.:72-43-5
- Description:Methoxychlor
- Formula:C₁₆H₁₅Cl₃O₂
- Description:Methoxychlor
: Accredited to ISO/IEC 17025.