Order Entry
Austria
ContactUsLinkComponent
 
Formate standard for ion chromatography, TraceCERT®, Supelco®
  44293-100ML
 :  Merck
 :  
undefined
Formate standard for ion chromatography, TraceCERT®, Supelco®
  44293-100ML
 :  Merck
 :  

 

  • Concentration:
    1000 mg/l
  • Matrix:
    H₂O
  • Pack type:
    HDPE bottle
  • Pk:
    100 mL
  • Description:
    Formate standard solution

 

 

Prepared with calcium formate and water, filtered through a 0,2 μm membrane filter and stabilised with sodium azide.

 : Certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034.