Order Entry
Austria
ContactUsLinkComponent
Calcium standard solution, for ion chromatography, TraceCERT®, Supelco®
Calcium standard solution, for ion chromatography, TraceCERT®, Supelco®
Catalog # 39865-100ML
Supplier:  Merck
CAS Number:  
undefined
Calcium standard solution, for ion chromatography, TraceCERT®, Supelco®
Catalog # 39865-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,1% HNO₃
  • Pack type:
    HDPE bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ300
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

About this item

Certifications: Certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034.