Order Entry
Austria
ContactUsLinkComponent
Calbiochem® Human recombinant interleukin-1 beta
Calbiochem® Human recombinant interleukin-1 beta
Catalog # 407615-5
CAS Number:  
Calbiochem® Human recombinant interleukin-1 beta
Catalog # 407615-5
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    –70 °C
  • Form:
    Lyophilised
  • Protein/peptide name:
    Interleukin-1 beta
  • Purity:
    >97% by SDS-PAGE
  • Endotoxin level:
    ≤1.0 EU/µg IL-1β

Specifications

About this item

Recombinant, human interleukin-1ß expressed in E. coli. Major form of interleukin-1 secreted by monocytes and macrophages. Induces nitric oxide synthase in pancreatic and smooth muscle cells and suppresses apoptosis in rat ovarian follicles. Involved in inflammatory and immune responses and promotes wound healing.