Order Entry
Austria
ContactUsLinkComponent
N,N-Dimethylacetamide ≥99.9% (by GC) for headspace gas chromatography
Irritant
Health
N,N-Dimethylacetamide ≥99.9% (by GC) for headspace gas chromatography
Catalog # APPC753145.1611
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Irritant
Health
N,N-Dimethylacetamide ≥99.9% (by GC) for headspace gas chromatography
Catalog # APPC753145.1611
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    1.000 mL
  • Seq:
    2865

Specifications

Specification Test Results

  • Minimum Assay (G.C.):
    99,9 %
  • Identity:
    IR Passes Test
  • Refractive Index n20/D:
    1.438 - 1.439
  • Apha Colour:
    ≤10
  • Suitability for Analysis of Residual Solvents Cg/Hs:
    ≤0.5 ppm
  • Solvents of Class 1 According To ICH:
    ≤5 ppm
  • Solvents of Class 2 According To ICH:
    ≤25 ppm
  • Solvents Of Class 3 According To ICH:
    ≤0.05%

Specification Test Results

About this item

 :
 :87.12 g/mol
Melting Pt:–20 °C
Flash Pt:70 °C
 :GHS07
 :GHS08
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686