VWR®, N-Plan Objectives
Supplier: VWR Collection
Jump to:
About this item
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
12 Options Available Below
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
VWR® for Microscopy
Product Literature
Product Family Options
Product Information
- MagnificationWorking distanceAvailabilityPrice
- 630-1639IOS N-plan achromatic objective
- 630-1640IOS N-plan achromatic objective
- 630-1641IOS N-plan achromatic objective
- 630-1642IOS N-plan achromatic objective
- 630-2049IOS N-plan achromatic objective
- 630-1643IOS N-plan achromatic objective (oil/water)IOS N-plan achromatic objective (oil/water)100×/ 1,25 mm (i)0,13 mmLog in for availability1 * 1 ST191,00 €
- 630-2004N-plan objective
- 630-2005N-plan objective
- 630-2006N-plan objective
- 630-2007N-plan objective
- 630-2008N-plan objective
- 630-2009N-plan objective (oil/water)