ContactUsLinkComponent
Radiometer Analytical XR110 Reference Electrode (Red Rod, Ground Joint, Screw Cap)
Catalog # HACHB20B112
Supplier:  Hach
undefined
Radiometer Analytical XR110 Reference Electrode (Red Rod, Ground Joint, Screw Cap)
Catalog # HACHB20B112
Supplier:  Hach
Specifications
  • Description:
    Reference electrode XR110, Hg free, 120 mm length
  • Material:
    Glass
  • Electrolyte:
    KCl 3 M
  • Pk:
    1
  • Temp.:
    −10...100 °C
Specifications
About this item
The XR110 reference electrode has a glass body, red rod Ag/AgCl reference technology system, refillable electrolyte (KCl 3 M), ground joint, and porous pin junction.
  • Recommended for voltammetry applications and educational purposes
The XR110 is a screw cap electrode and requires an external connection cable for connection to a meter.
Delivery: Delivery includes radiometer analytical XR110 reference electrode, 30 ml KCl 3M refill solution and operating instructions.