Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Registrieren
Products
Services
Digital Solutions
Applications
Support
About
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
Austria
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
ContactUsLinkComponent
Home
Services
Manufacturer's test certificate for multi-parameter meters
Manufacturer's test certificate for multi-parameter meters
Manufacturer's test certificate for multi-parameter meters
Artikelnr. 663-0107
Supplier:
SI Analytics
Expand image
Manufacturer's test certificate for multi-parameter meters
Artikelnr. 663-0107
Supplier:
SI Analytics
Specifications
Description:
Manufacturer's test certificate
Pk:
1
For:
pH, conductivity, oxygen and multi meters
Specifications
About this item
Product Details
Product Details