Orders LinkContactUsLinkComponent
Toggle drums
Toggle drums
Toggle drums
Supplier:  Sampling Systems
Certificates
Toggle drums
7 Options Available Below
Product Details & Documents
These stainless steel containers from 2 litres up to 300 litres are designed for storing and processing powders and liquids.
  • Splash-tight lid secured by toggle clamps
  • Hygienic solid rim
  • With two side handles
  • Fully welded, crevice free interior
  • Finished to a high quality mirror polish
  • Removable, easy clean silicone seal
Seal is food grade and meets all the latest FDA compliance standards.
Product Family Options
Product Information
  • Capacity
    Ø opening
    Height
    Availability
    Price
  • 2 L
    140 mm
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 556,00 €
    556,00 €
    1 * 1 ST
       556,00 €
  • 5 L
    196 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 689,00 €
    689,00 €
    1 * 1 ST
       689,00 €
  • 8 L
    196 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 822,00 €
    822,00 €
    1 * 1 ST
       822,00 €
  • 10 L
    230 mm
    270 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 944,00 €
    944,00 €
    1 * 1 ST
       944,00 €
  • 15 L
    230 mm
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €
  • 20 L
    300 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 090,00 €
    1 090,00 €
    1 * 1 ST
       1 090,00 €
  • 300 L
    710 mm
    800 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 450,00 €
    3 450,00 €
    1 * 1 ST
       3 450,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Silicone gasket (FDA acceptable), 300 mm int.Ø of drum
    Silicone gasket (FDA acceptable), 300 mm int.Ø of drum
    SAMPA430-300
    Log in for availability
    1 * 1 ST - 67,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Silicone gasket (FDA acceptable), 196 mm int.Ø of drum
    Silicone gasket (FDA acceptable), 196 mm int.Ø of drum
    SAMPA430-196
    Log in for availability
    1 * 1 ST - 45,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Drum lid, 316L SS, 470 mm int.Ø of drum
    Drum lid, 316L SS, 470 mm int.Ø of drum
    SAMPA429-470
    Log in for availability
    1 * 1 ST - 405,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Drum lid, 316L SS, 395 mm int.Ø of drum
    Drum lid, 316L SS, 395 mm int.Ø of drum
    SAMPA429-395
    Log in for availability
    1 * 1 ST - 396,00 €