Order Entry
Austria
ContactUsLinkComponent
Sigma-Aldrich® Intralipid, 20%, emulsion
Sigma-Aldrich® Intralipid, 20%, emulsion
Catalog # SIALI141-100ML
Supplier:  Merck
CAS Number:  
Sigma-Aldrich® Intralipid, 20%, emulsion
Catalog # SIALI141-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Intralipid, 20%

Specifications

About this item

Used as a solubilisation agent.

  • CAS: 68890-65-3
  • MDL: MFCD00287215

Application
Intralipid has been used:
for testing its optical properties to be used as scattering medium for tissue phantom generation
as a component of agarose gel phantom to reduce scatter coefficients and background absorption
in infusion into mice in hyperinsulinemic-euglycemic clamps studies to evaluate its effect on nonesterified free fatty acids (NEFA) circulation

Biochem/physiol Actions:
Intralipid is a lipid emulsion comprising soybean oil. It favors proinflammatory cytokines production and is recommended for the treatment of immune-mediated pregnancy and implantation failure. In parenteral nutrition, intralipid is the source of fats. It elicits protection in heart ischemia-reperfusion injury by inhibiting mitochondrial permeability transition pore opening. Intralipid is an effective alternative for intravenous immunoglobulin and improves bioavailability of platinum based anti-cancer drugs.