Order Entry
Austria
ContactUsLinkComponent
2-Methoxyestradiol ELISA kit
2-Methoxyestradiol ELISA kit
Supplier:  Cayman Chemical
Certificates
undefined
2-Methoxyestradiol ELISA kit
Supplier:  Cayman Chemical

Specifications

  • Assay Type:
    Competitive
  • Target protein:
    2-Methoxyestradiol
  • Developmental time:
    90 - 120 min
  • Mid point:
    330 - 390 pg/ml
  • Detection range:
    13,1 - 8000 pg/ml
  • Sensitivity:
    40 pg/ml
  • Incubation time:
    18 hr

Specifications

Related Information

Frequently Bought Together