Septa for screw caps, N 9
Supplier: MACHEREY-NAGEL
Certificates
About this item
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
(±)-1,2-Propanediol ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
No-Met biocompatible mobile phase filters
Supplier: VICI JOUR RESEARCH
Stoppers, rubber, conical
Supplier: KARTELL
Screw neck vials, N 9
Supplier: MACHEREY-NAGEL