Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Sieving tool

Sieving tool
  SAMPA516-120
Sieving tool
  SAMPA516-120
Sieving tool
Sieving tool

Sieving tool

 
282,00 €
   
1 * 1 ST
 

This stainless steel sieving tool is ideal for pushing powders through mesh screens. The heavy base can be used to help break down small lumps and agglomerates.

  • 316L Stainless steel
  • Radiused corners
  • Crevice free construction
  • Satin finish
  • Robust handle (150 mm length)

Overall height: 160 mm

Base (Ø×H):120×10 mm

Weight: 1,13 kg

 
  • Description:
    Sieving tool