Bovine Ribonuclease B (from Pancreas)
Supplier: Merck
Certificates
About this item
Ribonuclease B from bovine pancreas, purified by affinity chromatography is used in the digestion of RNA during cell cycle platform analysis.
Specifications
- CAS No.:9001-99-4
- Source:Pancreas
- Species:Bovine
- Storage conditions:−20 °C
- Enzyme name:Ribonuclease B
- Enzyme synonyms:RNase B
- Purity:≥80%
Specifications
Frequently Bought Together