Order Entry
Austria
ContactUsLinkComponent
 
Magnesium standard for IC, TraceCERT®, Supelco®
  89441-100ML
 :  Merck
 :  
undefined
Magnesium standard for IC, TraceCERT®, Supelco®
  89441-100ML
 :  Merck
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    0,1% HNO₃
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ300
  • Element:
    Magnesium

 

 

This standard is prepared with high purity Mg(NO₃)₂, HNO₃ and water.

 : Certified in accordance with ISO/IEC 17025 and ISO 17034