Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Vanadium Standards
Reference standard, vanadium, VHG, CONOSTAN
Reference standard, vanadium, VHG, CONOSTAN
Reference standard, vanadium, VHG, CONOSTAN
Catalog # PROOCON150-100-235
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
Reference standard, vanadium, VHG, CONOSTAN
Catalog # PROOCON150-100-235
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Concentration:
1000 µg/g
Matrix:
Base Oil 20
Pk:
50 g
Element:
Vanadium
Symbol:
V
Specifications
About this item
Product Details
Product Details