Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Food Standards
Organic reference standard, Wine - alcohol, ERM
Organic reference standard, Wine - alcohol, ERM
Organic reference standard, Wine - alcohol, ERM
Catalog # PROOERM-BA002
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
Organic reference standard, Wine - alcohol, ERM
Catalog # PROOERM-BA002
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Concentration:
10%
Pk:
1
Standards:
Reference standard
Description:
Wine - Alcohol
Pk Info:
250 ml
Specifications
About this item
Product Details
Product Details