ContactUsLinkComponent
Vector, pET-24(+) DNA
Vector, pET-24(+) DNA
Catalog # 69772-3
CAS Number:  
Vector, pET-24(+) DNA
Catalog # 69772-3
CAS Number:  
Specifications
  • Pk:
    10 µG
  • Description:
    Vector, pET-24(+) DNA, Novagen®
Specifications
About this item
pET-24(+) is a transcription vector designed for expression from bacterial translation signals carried within a cloned insert. It lacks the ribosome binding site and ATG start codon present on the pET translation vectors. A C-terminal His·Tag® sequence is available
Delivery: pET vectors are supplied as purified plasmid DNA (10 µg) and an induction control strain (supplied as a glycerol stock).