Vector, pET-24d(+) DNA
Catalog # 69752-3
Supplier: Merck Millipore (Novagen)
CAS Number:
Specifications
- Pk:10 µG
- Description:Vector, pET-24d(+) DNA, Novagen®
Specifications
About this item
The pET-24a-d(+) vectors carry an N-terminal T7·Tag® sequence plus an optional C-terminal His·Tag® sequence.
Delivery: The pET vectors are supplied as purified plasmid DNA (10 µg). And also includes an induction control strain (supplied as a glycerol stock).