InSolution™ MG-132
Lieferant: Merck Millipore (Calbiochem)
Zertifikate
Über diesen Artikel
InSolution™ MG-132 is provided as a 10 mM solution in DMSO. Acts as a potent, reversible proteasome inhibitor (Ki=4 nM).
- Packaged under inert gas
Spezifikationen
- CAS No.:133407-82-6
- Storage conditions:Stored at –20 °C
- Form:Liquid
- Protein/peptide name:MG-132
- Purity:>98% (by HPLC)
- Formulation:A 10 mM (1 mg/210 µl) or (5 mg/1050 µl) solution of MG-132 in DMSO
- Molecular formula:C₂₆H₄₁N₃O₅
Spezifikationen
Frequently Bought Together