Order Entry
Austria
ContactUsLinkComponent
Potassium chloride 99.0-100.5% (by argentometric analysis, on dry substance) USP, BP, Ph. Eur., pharma grade
Potassium chloride 99.0-100.5% (by argentometric analysis, on dry substance) USP, BP, Ph. Eur., pharma grade
Supplier:  PanReac AppliChem
Certificates
Potassium chloride 99.0-100.5% (by argentometric analysis, on dry substance) USP, BP, Ph. Eur., pharma grade
Supplier:  PanReac AppliChem

Specification Test Results

  • Assay (Argentometric):
    99.0 - 100.5%
  • Identity According to Pharmacopoeias:
    Passes Test
  • Acidity or Alkalinity:
    Passes Test
  • Loss on Drying at 105 °C:
    ≤1.0%
  • Bromide (Br):
    ≤0.1%
  • Sulphate (SO₄):
    ≤0.03%
  • Iodide:
    Passes Test
  • Residual Solvents (Ph.Eur./USP):
    Passes Test
  • Ba (Ph. Eur.):
    Passes Test
  • Calcium, Magnesium and Alkaline-Earth Metals (as Ca):
    ≤0.02%
  • Iodide (USP):
    ≤0.005%
  • Heavy Metals (as Pb):
    ≤0.001%
  • Al:
    ≤0.0001%
  • Ba:
    ≤0.001%
  • Fe:
    ≤0.002%
  • NA:
    ≤0.1%

Specification Test Results

Frequently Bought Together