
Sulphuric acid 75% for analysis
Catalog # APPC123374.1212
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay:Min. 75%
- Chloride:Max. 0.00001%
- Nitrate:Max. 0.00002%
- Phosphate:Max. 0.00005%
- Al:Max. 0.000005%
- As:Max. 0.000001%
- Ca:Max. 0.00005%
- Fe:Max. 0.00002%
- NA:Max. 0.00005%
- Ni:Max. 0.000002%
- PB:Max. 0.000002%
- Zn:Max. 0.000005%
Specification Test Results
About this item
:
:98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589