Order Entry
Austria
ContactUsLinkComponent
 
8-isoprostane express ELISA kit
 
undefined
8-isoprostane express ELISA kit

 

  • Assay Type:
    Competitive
  • Target protein:
    8-Isoprostane
  • Mid point:
    40 - 60 pg/ml (50% B/B₀)
  • Detection range:
    2,5 - 1500 pg/ml
  • Storage temperature:
    –20 °C
  • Sensitivity:
    10 pg/ml (80% B/B₀)

 

Frequently Bought Together