Shipping tubes, with screw cap
Supplier: SEMADENI
Certificates
About this item
PP or LDPE.
- Very stable and liquid-tight
- Excellent chemical and temperature resistance up to +130 °C
- Autoclavable (except for LDPE tube)
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together