Orders LinkContactUsLinkComponent
ICP multi-element standards (28 components)
 :   CPACHEM
ICP multi-element standards (28 components)
 :  CPACHEM
ICP multi-element standards (28 components)
ICP multi-element standards (28 components)
Product Family Options
  • Description
    Matrix
    Pk
  • ICP multi-element standard, 100 mg/l, (28 elements in in 5 % HNO₃) Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn
    5% HNO₃
    100 mL
    1 * 100 mL
    1 * 100 mL - 382,00 €
    382,00 €
    1 * 100 mL
       382,00 €
  • ICP multi-element standard, 100 mg/l, (28 elements in in 5 % HNO₃) Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn
    5% HNO₃
    500 mL
    1 * 500 mL
    1 * 500 mL - 799,00 €
    799,00 €
    1 * 500 mL
       799,00 €