- Pk:1 KIT
- Description:NF-κB (p65) Transcription Factor Assay Kit, 96 strip wells
- Storage temperature:−80 °C
NF-κB (p65) Transcription Factor Assay Kit is a 96-well assay for measurement of NF-κB (p65).
- A sensitive, non-radioactive method of detecting NF-κB (p65) from whole cell lysates
- 96-well plate format replaces EMSAs
- Capture the transcription factor using a specific dsDNA sequence bound to the plate
- Detect the dsDNA-bound transcription factor with specific antibodies in an ELISA format
Cayman’s NF-κB (p65) Transcription Factor Assay is a non-radioactive, sensitive method for detecting specific transcription factor DNA binding activity in nuclear extracts and whole cell lysates. A 96-well enzyme-linked immunosorbent assay (ELISA) replaces the cumbersome radioactive electrophoretic mobility shift assay (EMSA). A specific double stranded DNA (dsDNA) sequence containing the NF-κB response element is immobilized to the wells of a 96-well plate. NF-κB contained in a nuclear extract, binds specifically to the NF-κB response element. NF-κB (p65) is detected by addition of specific primary antibody directed against NF-κB (p65). A secondary antibody conjugated to HRP is added to provide a sensitive colorimetric readout at 450 nm. Cayman’s NF-κB (p65) Transcription Factor Assay detects human NF-κB (p65). It will not cross-react with NF-κB (p50).