Order Entry
Austria
ContactUsLinkComponent
 
Multi-element ion chromatography anion standard solution, CRM, Supelco®
Catalog # 89886-50ML-F
Supplier:  Merck
CAS Number:  
Multi-element ion chromatography anion standard solution, CRM, Supelco®
Catalog # 89886-50ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10,0 mg/kg ± 0,2
  • Pk:
    50 mL
  • Description:
    Multi-anion standard, (F⁻, Cl⁻, Br⁻, NO₃⁻, PO₄⁻³, SO₄⁻²)
  • M-Clarity quality level:
    MQ300

Specifications

About this item

This standard solution is a certified primary reference material for ion chromatography.

Produced with certified high purity materials (NaF, NaCl, NaBr, NaNO₃, Na₂HPO₄, Na₂SO₄) and ultrapure water (18 MOhm, 0,22 μm filtered).

Certifications: Traceable to SI.