Mouse IgM purification kit
Supplier: LIGATRAP TECHNOLOGIES
Certificates
About this item
LigaTrap® Mouse IgM Purification Kit with LT technology utilises novel, patented ligands, designed for IgM antibody purification. Kit provides all the necessary reagents for fast, convenient micro-scale purification of mouse IgM in just ten easy steps.
- Designed to purify mouse IgM
- For in vitro laboratory use
- Binding capacity for mouse IgM is >10 mg/ml resin
- Storage: 2 to 8 °C
LigaTrap® mouse IgM purification resin is engineered to purify high quality mouse IgM antibodies from recombinant and monoclonal sources. Resins bind an exceptionally broad range of recombinant, monoclonal and human/animal source polyclonal immunoglobulins.
Each spin column can be used, regenerated, and used up to ten times with minimal
loss in binding capacity.
Delivery: Each kit contains ten microspin columns prefilled with 0,1 ml of LigaTrap® mouse IgM purification resin, buffers and collection tubes.
Caution: Not for diagnostic or therapeutic use. Not for human or animal consumption.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together