ContactUsLinkComponent
TBX (Tryptone Bile X‑glucuronide) agar, Chromocult®
TBX (Tryptone Bile X‑glucuronide) agar, Chromocult®
Catalog # 1.16122.0500
Supplier:  Merck
CAS Number:  
undefined
TBX (Tryptone Bile X‑glucuronide) agar, Chromocult®
Catalog # 1.16122.0500
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    500 g
  • Standards:
    ISO 16649
  • Description:
    Chromocult® TBX (Tryptone bile agar)
Specifications
About this item
For the enumeration, detection and identification of β-glucuronidase positive Escherichia coli from food and feeding stuff as well as from environmental samples in the area of food production and food handling.
Certifications: Complies with the specifications given by ISO 16649 part 1 and part 2 and with those given by EN ISO 16649-3.