Ferroin (1,10-phenanthrolineferrous sulphate complex) solution Reag. Ph. Eur. redox indicator E0 (H2SO4 1 mol/l) = +1,06 volt, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:14634-91-4
:
Formula:C₃₆H₂₄FeN₆SO₄
MDL Number:MFCD00036428
:692.54 g/mol
Boiling Pt:100 °C (1.013 hPa)
Density:1.00 g/cm³ (20 °C)
Storage Temperature:Ambient
Specification Test Results
- Change:complying
Specification Test Results
Frequently Bought Together